Assay Details
Target Gene Details
Entrez Gene ID: | 60436 |
Gene Name: | TGFB induced factor homeobox 2 |
Gene Aliases: |
- |
Location: |
Chr.20:36573473-36593952 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGIF2 | NM_001199513.1 | NP_001186442.1 | ||
NM_001199514.1 | NP_001186443.1 | |||
NM_001199515.1 | NP_001186444.1 | |||
NM_021809.6 | NP_068581.1 | |||
AB042646.1 | BAB16424.1 | |||
AK074802.1 | BAC11216.1 | |||
AK313919.1 | ||||
AL705108.1 | ||||
BC006549.2 | AAH06549.1 | |||
BC012816.2 | AAH12816.1 | |||
BG723865.1 | ||||
BP353213.1 | ||||
DA438831.1 |
Target Gene Details
Entrez Gene ID: | 100527943 |
Gene Name: | TGIF2-C20orf24 readthrough |
Gene Aliases: |
- |
Location: |
Chr.20:36574554-36612557 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGIF2-C20orf24 | NM_001199535.1 | NP_001186464.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833967 | Chr.20:36477427 - 36658869 on Build GRCh38 | Loss | TGIF2-C20orf24 TGIF2 C20orf24 NDRG3 DLGAP4-AS1 MYL9 SLA2 DLGAP4 |
nsv1057192 | Chr.20:36514051 - 36839310 on Build GRCh38 | Loss | TGIF2-C20orf24 TGIF2 C20orf24 NDRG3 DLGAP4-AS1 MYL9 SLA2 DLGAP4 SOGA1 DSN1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |