Assay Details
Target Gene Details
Entrez Gene ID: | 100130264 |
Gene Name: | uncharacterized LOC100130264 |
Gene Aliases: |
- |
Location: |
Chr.20:19242302-19284596 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100130264 | NR_024564.1 | |||
AK097497.1 | BAC05077.1 |
Target Gene Details
Entrez Gene ID: | 57419 |
Gene Name: | solute carrier family 24 member 3 |
Gene Aliases: |
NCKX3 |
Location: |
Chr.20:19212646-19722897 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC24A3 | NM_020689.3 | NP_065740.2 | ||
AF169257.2 | AAG12988.2 | |||
AF288087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv585671 | Chr.20:19231033 - 19273047 on Build GRCh38 | Loss | SLC24A3 LOC100130264 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |