Assay Details
Target Gene Details
Entrez Gene ID: | 79913 |
Gene Name: | ARP5 actin-related protein 5 homolog |
Gene Aliases: |
Arp5, INO80M |
Location: |
Chr.20:38748454-38772446 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ACTR5 | NM_024855.3 | NP_079131.3 | ||
AK022847.1 | BAB14270.1 | |||
BC001324.1 | AAH01324.1 | |||
BC006162.1 | AAH06162.1 | |||
BC038402.1 | AAH38402.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1058457 | Chr.20:38277492 - 38844429 on Build GRCh38 | Loss | ARHGAP40 BPI ACTR5 ADIG SNORA71D SNHG11 SLC32A1 SNORA71A LBP RALGAPB SNORA71B SNORA71E SNORA60 SNHG17 LOC149684 SNORA71C MIR548O2 PPP1R16B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |