Assay Details
Target Gene Details
Entrez Gene ID: | 128869 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class U |
Gene Aliases: |
CDC91L1, GAB1 |
Location: |
Chr.20:34560542-34677285 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIGU | NM_080476.4 | NP_536724.1 | ||
XM_011528542.2 | XP_011526844.1 | |||
XM_017027664.1 | XP_016883153.1 | |||
AB086842.1 | BAC53626.1 | |||
AK075507.1 | BAC11660.1 | |||
AK301900.1 | ||||
AY339061.1 | AAQ18022.1 | |||
AY358816.1 | AAQ89175.1 | |||
AY422169.1 | AAR23798.1 | |||
BC030512.1 | AAH30512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv3359 | Chr.20:34531574 - 34576232 on Build GRCh38 | Deletion | PIGU MAP1LC3A DYNLRB1 |
nsv3360 | Chr.20:34562312 - 34597446 on Build GRCh38 | Insertion | PIGU |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |