Assay Details
Target Gene Details
Entrez Gene ID: | 10406 |
Gene Name: | WAP four-disulfide core domain 2 |
Gene Aliases: |
EDDM4, HE4, WAP5, dJ461P17.6 |
Location: |
Chr.20:45469754-45481532 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
WFDC2 | NM_006103.3 | NP_006094.3 | ||
AF330259.1 | AAL37485.1 | |||
AF330260.1 | AAL37486.1 | |||
AF330262.1 | AAL37488.1 | |||
AK225528.1 | ||||
AK311966.1 | ||||
AY212888.1 | AAO52683.1 | |||
BC046106.1 | AAH46106.1 | |||
BI759775.1 | ||||
CR456977.1 | CAG33258.1 | |||
X63187.1 | CAA44869.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss | WFDC2 KCNS1 RBPJL SYS1-DBNDD2 WFDC5 STK4 RIMS4 SLPI WFDC8 PI3 KCNK15 PIGT PABPC1L SYS1 EPPIN-WFDC6 KCNK15-AS1 MATN4 SPINT3 MIR6812 WFDC12 SDC4 WFDC10A SEMG2 DBNDD2 WFDC9 YWHAB TOMM34 WFDC6 STK4-AS1 TP53TG5 WISP2 SEMG1 EPPIN |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |