Assay Details
Target Gene Details
Entrez Gene ID: | 26039 |
Gene Name: | SS18L1, nBAF chromatin remodeling complex subunit |
Gene Aliases: |
CREST, LP2261 |
Location: |
Chr.20:62143720-62182510 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SS18L1 | NM_001301778.1 | NP_001288707.1 | ||
NM_198935.2 | NP_945173.1 | |||
NR_125980.1 | ||||
NR_125981.1 | ||||
XM_005260389.3 | XP_005260446.1 | |||
XM_005260391.1 | XP_005260448.1 | |||
XM_017027784.1 | XP_016883273.1 | |||
AB014593.1 | ||||
AK125656.1 | ||||
AK291485.1 | ||||
AK299881.1 | ||||
AK308765.1 | ||||
BX640848.1 | CAE45918.1 | |||
DC375156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509782 | Chr.20:62125636 - 62180328 on Build GRCh38 | Insertion | SS18L1 PSMA7 LSM14B |
nsv586481 | Chr.20:62143364 - 62158583 on Build GRCh38 | Gain | SS18L1 PSMA7 |
nsv3440 | Chr.20:62137705 - 62169472 on Build GRCh38 | Insertion | SS18L1 PSMA7 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |