Assay Details
Target Gene Details
Entrez Gene ID: | 54363 |
Gene Name: | hydroxyacid oxidase 1 |
Gene Aliases: |
GOX, GOX1, HAOX1 |
Location: |
Chr.20:7882984-7940446 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAO1 | NM_017545.2 | NP_060015.1 | ||
AB024079.1 | BAA82872.1 | |||
AF231916.1 | AAF40199.1 | |||
AF244134.1 | AAF63219.1 | |||
AK289423.1 | ||||
AK308963.1 | ||||
AK313839.1 | ||||
AL121739.1 | CAB57329.1 | |||
BC113665.1 | ||||
BC113667.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1059059 | Chr.20:7793987 - 8053708 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv585384 | Chr.20:7571617 - 8336371 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv3893279 | Chr.20:7582024 - 7999743 on Build GRCh38 | Gain |
![]() ![]() |
nsv833915 | Chr.20:7798839 - 7957303 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)