Assay Details
Target Gene Details
Entrez Gene ID: | 388799 |
Gene Name: | family with sequence similarity 209 member B |
Gene Aliases: |
C20orf107, dJ1153D9.4 |
Location: |
Chr.20:56527124-56555626 on Build GRCh38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM209B |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv525425 | Chr.20:56403979 - 56897294 on Build GRCh38 | Gain | FAM209A LOC105372682 TFAP2C GCNT7 RTFDC1 CSTF1 CASS4 FAM209B |
nsv834015 | Chr.20:56518712 - 56656502 on Build GRCh38 | Loss | FAM209A LOC105372682 TFAP2C GCNT7 RTFDC1 FAM209B |
nsv1056574 | Chr.20:56495687 - 56761211 on Build GRCh38 | Gain | FAM209A LOC105372682 TFAP2C GCNT7 RTFDC1 FAM209B |