Assay Details
Target Gene Details
Entrez Gene ID: | 22888 |
Gene Name: | U-box domain containing 5 |
Gene Aliases: |
RNF37, UBCE7IP5, UIP5, hUIP5 |
Location: |
Chr.20:3107573-3159910 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBOX5 | NM_001267584.1 | NP_001254513.1 | ||
NM_014948.3 | NP_055763.1 | |||
NM_199415.2 | NP_955447.1 | |||
AB020667.1 | BAA74883.2 | |||
AK022444.1 | ||||
AK222570.1 | BAD96290.1 | |||
AK222876.1 | BAD96596.1 | |||
BC000515.2 | AAH00515.1 | |||
BC046122.1 | AAH46122.1 | |||
CR457089.1 | CAG33370.1 |
Target Gene Details
Entrez Gene ID: | 100134015 |
Gene Name: | UBOX5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:3106911-3150867 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UBOX5-AS1 | NR_038395.1 | |||
AK097516.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv585276 | Chr.20:3030542 - 3366307 on Build GRCh38 | Gain | MRPS26 FASTKD5 OXT DDRGK1 GNRH2 UBOX5-AS1 ITPA SLC4A11 UBOX5 PTPRA LZTS3 AVP C20orf194 |
nsv833897 | Chr.20:2990324 - 3175146 on Build GRCh38 | Loss | MRPS26 FASTKD5 UBOX5 OXT PTPRA LZTS3 GNRH2 UBOX5-AS1 AVP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |