Assay Details
Target Gene Details
Entrez Gene ID: | 128866 |
Gene Name: | charged multivesicular body protein 4B |
Gene Aliases: |
C20orf178, CHMP4A, CTPP3, CTRCT31, SNF7, SNF7-2, Shax1, VPS32B, Vps32-2, dJ553F4.4 |
Location: |
Chr.20:33811304-33854367 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHMP4B | NM_176812.4 | NP_789782.1 | ||
AB100261.1 | BAC79375.1 | |||
AY329085.1 | AAQ91194.1 | |||
AY726594.1 | ||||
BC033859.1 | AAH33859.1 | |||
CT001913.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv585930 | Chr.20:33779897 - 33839036 on Build GRCh38 | Gain | CHMP4B ZNF341 ZNF341-AS1 |
nsv518926 | Chr.20:33741162 - 33902088 on Build GRCh38 | Gain | CHMP4B ZNF341 ZNF341-AS1 |
nsv833961 | Chr.20:33605380 - 33836992 on Build GRCh38 | Loss | C20orf144 PXMP4 CHMP4B ACTL10 ZNF341 CBFA2T2 E2F1 NECAB3 ZNF341-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |