Assay Details
Target Gene Details
Entrez Gene ID: | 26090 |
Gene Name: | abhydrolase domain containing 12 |
Gene Aliases: |
ABHD12A, BEM46L2, C20orf22, PHARC, dJ965G21.2 |
Location: |
Chr.20:25294743-25390982 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ABHD12 | NM_015600.4 | NP_056415.1 | ||
BC014049.2 | AAH14049.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv779e214 | Chr.20:25104610 - 25367702 on Build GRCh38 | Gain | LOC105372580 LOC284798 PYGB LOC101926889 ABHD12 ENTPD6 |
nsv522873 | Chr.20:23923336 - 26134874 on Build GRCh38 | Gain | FAM182B FLJ33581 ZNF337 PYGB NINL NCOR1P1 NANP APMAP FAM182A LOC100134868 LOC284798 ZNF337-AS1 LOC101926955 ACSS1 LOC101926889 VN1R108P SYNDIG1 LOC105372582 LOC105372580 GINS1 ENTPD6 LOC105372585 GGTLC1 ABHD12 CST7 VSX1 LOC101926935 |
nsv585746 | Chr.20:25090818 - 25489420 on Build GRCh38 | Gain | LOC105372580 LOC284798 PYGB NINL GINS1 LOC101926889 ABHD12 ENTPD6 |
dgv4290n100 | Chr.20:25086650 - 25494993 on Build GRCh38 | Gain | LOC105372580 LOC284798 PYGB NINL GINS1 LOC101926889 ABHD12 ENTPD6 |
esv3645582 | Chr.20:25284710 - 25318462 on Build GRCh38 | Loss | PYGB ABHD12 |
nsv833948 | Chr.20:25251090 - 25429904 on Build GRCh38 | Loss | PYGB GINS1 ABHD12 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |