Assay Details
Target Gene Details
Entrez Gene ID: | 1591 |
Gene Name: | cytochrome P450 family 24 subfamily A member 1 |
Gene Aliases: |
CP24, CYP24, HCAI, HCINF1, P450-CC24 |
Location: |
Chr.20:54145731-54173985 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CYP24A1 | XM_017027692.1 | XP_016883181.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv586224 | Chr.20:53858311 - 54662951 on Build GRCh38 | Gain | PFDN4 CYP24A1 MIR4756 BCAS1 DOK5 SUMO1P1 |
nsv834010 | Chr.20:54143285 - 54316400 on Build GRCh38 | Loss | PFDN4 CYP24A1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |