Assay Details
Target Gene Details
Entrez Gene ID: | 30813 |
Gene Name: | visual system homeobox 1 |
Gene Aliases: |
CAASDS, KTCN, KTCN1, PPCD, PPCD1, PPD, RINX |
Location: |
Chr.20:25069940-25082379 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
VSX1 | NM_001256271.1 | NP_001243200.1 | ||
NM_001256272.1 | NP_001243201.1 | |||
NR_045948.1 | ||||
NR_045951.1 | ||||
XM_017027837.1 | XP_016883326.1 | |||
XM_017027838.1 | XP_016883327.1 | |||
DQ854809.1 | ||||
DQ854810.1 | ||||
DQ854811.1 | ||||
DQ854812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1056345 | Chr.20:24838934 - 25187523 on Build GRCh38 | Gain | CST7 APMAP VSX1 ACSS1 LOC284798 LOC101926889 |
esv3645552 | Chr.20:24494271 - 25093181 on Build GRCh38 | Gain | CST7 APMAP VSX1 ACSS1 SYNDIG1 |
nsv522873 | Chr.20:23923336 - 26134874 on Build GRCh38 | Gain | LOC105372580 FAM182A LOC105372585 ACSS1 LOC100134868 FLJ33581 VSX1 GGTLC1 GINS1 ZNF337 LOC101926889 FAM182B CST7 NANP APMAP SYNDIG1 ZNF337-AS1 ABHD12 LOC105372582 NINL LOC284798 LOC101926955 VN1R108P NCOR1P1 PYGB LOC101926935 ENTPD6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |