Assay Details
Target Gene Details
Entrez Gene ID: | 101927631 |
Gene Name: | ADNP antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:50930984-50945134 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADNP-AS1 | NR_110007.1 | |||
NR_110008.1 | ||||
NR_110009.1 | ||||
AI827739.1 | ||||
BF064247.1 | ||||
HY015168.1 |
Target Gene Details
Entrez Gene ID: | 8813 |
Gene Name: | dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit |
Gene Aliases: |
CDGIE, MPDS |
Location: |
Chr.20:50934855-50958564 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DPM1 | NM_001317034.1 | NP_001303963.1 | ||
NM_001317035.1 | NP_001303964.1 | |||
NM_001317036.1 | NP_001303965.1 | |||
NM_003859.2 | NP_003850.1 | |||
NR_133648.1 | ||||
AF007875.1 | AAC98797.1 | |||
AK289569.1 | ||||
BC007073.1 | AAH07073.1 | |||
BC008466.1 | AAH08466.1 | |||
BC016322.1 | AAH16322.1 | |||
BM922866.1 | ||||
BQ441542.1 | ||||
CA865473.1 | ||||
CR456926.1 | CAG33207.1 | |||
D86198.1 | BAA25646.1 | |||
DR156925.1 | ||||
KU178444.1 | ||||
KU178445.1 | ||||
KU178446.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv834002 | Chr.20:50810235 - 50986764 on Build GRCh38 | Loss | BCAS4 TMSB4XP6 DPM1 ADNP-AS1 ADNP MOCS3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |