Assay Details
Target Gene Details
Entrez Gene ID: | 101929225 |
Gene Name: | MCM8 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:5990795-6005821 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MCM8-AS1 | NR_110101.1 | |||
AK058185.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3575356 | Chr.20:5954200 - 6068502 on Build GRCh38 | Gain | LRRN4 MCM8-AS1 CRLS1 MCM8 |