Assay Details
Target Gene Details
Entrez Gene ID: | 100533975 |
Gene Name: | SLMO2-ATP5E readthrough |
Gene Aliases: |
- |
Location: |
Chr.20:59028678-59042846 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLMO2-ATP5E | NR_037929.1 | |||
NR_037930.1 | ||||
CD522026.1 | ||||
FY212904.1 |