Assay Details
Target Gene Details
Entrez Gene ID: | 100505622 |
Gene Name: | uncharacterized LOC100505622 |
Gene Aliases: |
- |
Location: |
Chr.19:42132629-42137099 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC100505622 | NR_038332.1 |
Target Gene Details
Entrez Gene ID: | 5452 |
Gene Name: | POU class 2 homeobox 2 |
Gene Aliases: |
OCT2, OTF2, Oct-2 |
Location: |
Chr.19:42086110-42197931 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POU2F2 | XM_017026886.1 | XP_016882375.1 | ||
XM_017026887.1 | XP_016882376.1 | |||
XM_017026888.1 | XP_016882377.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | LIPE-AS1 MIR4323 ATP1A3 LOC100505622 MIR8077 RABAC1 MIR6797 ZNF526 ERF LOC390937 PRR19 LOC101930071 PAFAH1B3 CNFN ARHGEF1 LIPE RPS19 TMEM145 GRIK5 MEGF8 CIC CD79A LOC100505585 POU2F2 DEDD2 ZNF574 GSK3A |
nsv2496 | Chr.19:42130710 - 42165604 on Build GRCh38 | Insertion | MIR4323 LOC100505622 POU2F2 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | LIPE-AS1 MIR4323 ATP1A3 LOC100505622 MIR8077 CEACAM4 RABAC1 CEACAM3 MIR6797 ZNF526 ERF LYPD4 LOC390937 CEACAM7 PRR19 LOC101930071 PAFAH1B3 CNFN ARHGEF1 CEACAM5 DMRTC2 CEACAM6 LIPE RPS19 TMEM145 GRIK5 MEGF8 CIC CD79A LOC100505585 POU2F2 DEDD2 ZNF574 GSK3A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |