Assay Details
Target Gene Details
Entrez Gene ID: | 10036 |
Gene Name: | chromatin assembly factor 1 subunit A |
Gene Aliases: |
CAF-1, CAF1, CAF1B, CAF1P150, P150 |
Location: |
Chr.19:4402596-4448322 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHAF1A | NM_005483.2 | NP_005474.2 | ||
XM_011527605.2 | XP_011525907.1 | |||
XM_011527606.2 | XP_011525908.1 | |||
XM_011527607.2 | XP_011525909.1 | |||
BC052620.1 | AAH52620.1 | |||
BC067093.1 | AAH67093.1 | |||
U20979.1 | AAA76736.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833720 | Chr.19:4367721 - 4540556 on Build GRCh38 | Loss | HDGFRP2 CHAF1A SH3GL1 PLIN4 LRG1 PLIN5 MIR4746 UBXN6 |
nsv833718 | Chr.19:4221577 - 4480248 on Build GRCh38 | Loss | ANKRD24 SH3GL1 EBI3 STAP2 MIR4746 UBXN6 CCDC94 HDGFRP2 FSD1 CHAF1A SHD TMIGD2 MPND |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |