Assay Details
Target Gene Details
Entrez Gene ID: | 558 |
Gene Name: | AXL receptor tyrosine kinase |
Gene Aliases: |
ARK, JTK11, Tyro7, UFO |
Location: |
Chr.19:41219199-41261767 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AXL | NM_001278599.1 | NP_001265528.1 | ||
NM_001699.5 | NP_001690.2 | |||
NM_021913.4 | NP_068713.2 | |||
AK309139.1 | ||||
AK310881.1 | ||||
BC032229.1 | AAH32229.1 | |||
M76125.1 | AAA61243.1 | |||
S65125.1 | AAB20305.1 | |||
X57019.1 | CAA40338.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Additional Information:
For this assay, SNP(s) [rs80197793] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)