Assay Details
Target Gene Details
Entrez Gene ID: | 199786 |
Gene Name: | family with sequence similarity 129 member C |
Gene Aliases: |
BCNP1 |
Location: |
Chr.19:17523301-17556755 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FAM129C | NM_001098524.1 | NP_001091994.1 | ||
NM_173544.4 | NP_775815.2 | |||
AY254197.1 | AAO83574.1 | |||
AY254198.1 | AAO83575.1 | |||
AY254199.1 | AAO83576.1 |