Assay Details
Target Gene Details
Entrez Gene ID: | 147923 |
Gene Name: | zinc finger protein 420 |
Gene Aliases: |
APAK |
Location: |
Chr.19:37007815-37130322 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF420 | XM_011526503.2 | XP_011524805.1 | ||
XM_011526510.2 | XP_011524812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv960832 | Chr.19:37028939 - 37062279 on Build GRCh38 | Duplication | ZNF420 |
nsv579461 | Chr.19:36834924 - 37256206 on Build GRCh38 | Gain | ZNF585B ZNF790 ZNF383 ZNF345 LINC01535 ZNF585A ZNF568 ZNF420 ZNF829 |
esv3893201 | Chr.19:36973473 - 37170023 on Build GRCh38 | Gain | ZNF585A ZNF568 ZNF420 |
nsv833818 | Chr.19:36936906 - 37111898 on Build GRCh38 | Gain | ZNF568 ZNF420 |
nsv1064851 | Chr.19:36419357 - 37198819 on Build GRCh38 | Loss | ZNF790 LOC728752 ZNF461 ZNF420 ZNF382 ZNF260 LINC01534 ZNF585B ZNF850 ZNF345 LOC728485 ZNF585A ZNF567 ZNF568 ZNF529 LOC644189 ZNF566 ZNF790-AS1 ZNF829 ZNF529-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |