Assay Details
Target Gene Details
Entrez Gene ID: | 4784 |
Gene Name: | nuclear factor I X |
Gene Aliases: |
MRSHSS, NF1A, SOTOS2 |
Location: |
Chr.19:12995512-13098796 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFIX | NM_002501.3 | NP_002492.2 | ||
XM_005259918.4 | XP_005259975.1 | |||
XM_017026837.1 | XP_016882326.1 | |||
BF115883.1 | ||||
BT019732.1 | AAV38537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv828444 | Chr.19:13012681 - 13027353 on Build GRCh38 | Loss | NFIX |
nsv1055900 | Chr.19:12979147 - 13034197 on Build GRCh38 | Gain | NFIX |
nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion | RNASEH2A FARSA PRDX2 JUNB DNASE2 GCDH MIR5684 RTBDN RAD23A SYCE2 KLF1 NFIX CALR GADD45GIP1 SNORD41 MIR6794 MIR6515 MAST1 DHPS C19orf43 LOC105372280 WDR83OS TNPO2 MIR5695 WDR83 ASNA1 BEST2 SNORD135 DAND5 HOOK2 FBXW9 |
nsv2422 | Chr.19:13000362 - 13032711 on Build GRCh38 | Insertion | NFIX |
nsv833754 | Chr.19:12876208 - 13028186 on Build GRCh38 | Loss | MIR5695 GADD45GIP1 FARSA MIR6515 RAD23A DAND5 DNASE2 SYCE2 GCDH KLF1 NFIX CALR |
esv275430 | Chr.19:13015145 - 13020565 on Build GRCh38 | Gain+Loss | NFIX |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |