Assay Details
Target Gene Details
Entrez Gene ID: | 147923 |
Gene Name: | zinc finger protein 420 |
Gene Aliases: |
APAK |
Location: |
Chr.19:37007815-37130322 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF420 | XM_011526503.2 | XP_011524805.1 | ||
XM_011526510.2 | XP_011524812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv509740 | Chr.19:36970878 - 37041708 on Build GRCh38 | Insertion | ZNF420 ZNF568 |
nsv579461 | Chr.19:36834924 - 37256206 on Build GRCh38 | Gain | LINC01535 ZNF585A ZNF420 ZNF585B ZNF568 ZNF345 ZNF790 ZNF829 ZNF383 |
esv3644276 | Chr.19:37023082 - 37025753 on Build GRCh38 | Loss | ZNF420 |
esv3893201 | Chr.19:36973473 - 37170023 on Build GRCh38 | Gain | ZNF585A ZNF420 ZNF568 |
nsv833818 | Chr.19:36936906 - 37111898 on Build GRCh38 | Gain | ZNF420 ZNF568 |
nsv1064851 | Chr.19:36419357 - 37198819 on Build GRCh38 | Loss | LOC644189 ZNF566 ZNF461 ZNF420 ZNF585B ZNF568 ZNF345 ZNF260 LOC728752 LOC728485 ZNF790-AS1 ZNF567 LINC01534 ZNF529-AS1 ZNF585A ZNF382 ZNF790 ZNF829 ZNF850 ZNF529 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |