Assay Details
Target Gene Details
Entrez Gene ID: | 64763 |
Gene Name: | zinc finger protein 574 |
Gene Aliases: |
FP972 |
Location: |
Chr.19:42068477-42081567 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ZNF574 | XM_017027149.1 | XP_016882638.1 | ||
AK131369.1 | BAD18520.1 | |||
AK131418.1 | BAD18565.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv520239 | Chr.19:41853387 - 42426852 on Build GRCh38 | Loss | PRR19 CNFN ATP1A3 MIR6797 RABAC1 TMEM145 RPS19 LIPE MEGF8 CIC MIR8077 ZNF526 LIPE-AS1 PAFAH1B3 LOC100505622 POU2F2 LOC101930071 GSK3A GRIK5 CD79A MIR4323 ERF DEDD2 LOC100505585 ARHGEF1 LOC390937 ZNF574 |
nsv522854 | Chr.19:41613927 - 42426852 on Build GRCh38 | Gain | PRR19 CNFN ATP1A3 MIR6797 RABAC1 CEACAM6 TMEM145 CEACAM5 RPS19 LIPE MEGF8 CIC MIR8077 ZNF526 LIPE-AS1 PAFAH1B3 CEACAM4 LYPD4 LOC100505622 POU2F2 LOC101930071 GSK3A GRIK5 CD79A MIR4323 ERF DEDD2 CEACAM7 DMRTC2 LOC100505585 CEACAM3 ARHGEF1 LOC390937 ZNF574 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |