Assay Details
Target Gene Details
Entrez Gene ID: | 3609 |
Gene Name: | interleukin enhancer binding factor 3 |
Gene Aliases: |
CBTF, DRBF, DRBP76, MMP4, MPHOSPH4, MPP4, NF-AT-90, NF110, NF110b, NF90, NF90a, NF90b, NFAR, NFAR-1, NFAR2, TCP110, TCP80 |
Location: |
Chr.19:10654261-10692419 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ILF3 | NM_001137673.1 | NP_001131145.1 | ||
NM_004516.3 | NP_004507.2 | |||
NM_012218.3 | NP_036350.2 | |||
NM_017620.2 | NP_060090.2 | |||
NM_153464.2 | NP_703194.1 | |||
XM_006722742.2 | XP_006722805.1 | |||
XM_011527984.2 | XP_011526286.2 | |||
XM_011527985.2 | XP_011526287.1 | |||
XM_017026763.1 | XP_016882252.1 | |||
XM_017026765.1 | XP_016882254.1 | |||
AB209062.1 | BAD92299.1 | |||
AF141870.1 | AAD37575.1 | |||
AK291205.1 | ||||
AK291617.1 | ||||
AK293170.1 | ||||
AK294084.1 | ||||
BC048314.1 | AAH48314.1 | |||
BC064836.1 | AAH64836.1 | |||
DC320902.1 | ||||
U10324.1 | AAA20994.1 | |||
X98264.1 | CAA66917.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1160584 | Chr.19:10618877 - 10892186 on Build GRCh38 | Deletion | ILF3-AS1 QTRT1 MIR199A1 SLC44A2 CARM1 MIR4748 MIR638 TMED1 ILF3 MIR6793 C19orf38 DNM2 |
nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss | ILF3-AS1 QTRT1 SLC44A2 CDKN2D MIR638 ATG4D ILF3 MIR1238 KRI1 AP1M2 DNM2 |
esv3643657 | Chr.19:10657548 - 10661097 on Build GRCh38 | Loss | ILF3 |
esv1002882 | Chr.19:10654865 - 10660243 on Build GRCh38 | Deletion | ILF3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |