Assay Details
Target Gene Details
Entrez Gene ID: | 56301 |
Gene Name: | solute carrier family 7 member 10 |
Gene Aliases: |
ASC1, HASC-1, asc-1 |
Location: |
Chr.19:33208664-33225850 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC7A10 | NM_019849.2 | NP_062823.1 | ||
XM_006723284.2 | XP_006723347.1 | |||
XM_011527119.1 | XP_011525421.1 | |||
XM_011527120.1 | XP_011525422.1 | |||
AB037670.1 | BAB03213.1 | |||
AJ277731.1 | CAC81900.1 | |||
AK307527.1 | ||||
AK316594.1 | ||||
BC035627.1 | AAH35627.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833808 | Chr.19:33130887 - 33350702 on Build GRCh38 | Loss | LRP3 WDR88 CEBPA SLC7A10 CEBPA-AS1 |
nsv953277 | Chr.19:33192995 - 33270394 on Build GRCh38 | Deletion | LRP3 SLC7A10 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |