Assay Details
Target Gene Details
Entrez Gene ID: | 55655 |
Gene Name: | NLR family pyrin domain containing 2 |
Gene Aliases: |
CLR19.9, NALP2, NBS1, PAN1, PYPAF2 |
Location: |
Chr.19:54965284-55001142 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NLRP2 | NM_001174081.1 | NP_001167552.1 | ||
NM_001174082.1 | NP_001167553.1 | |||
NM_001174083.1 | NP_001167554.1 | |||
NM_017852.3 | NP_060322.1 | |||
AB209300.1 | BAD92537.1 | |||
AF298547.1 | AAG15253.1 | |||
AF310106.1 | AAG30289.1 | |||
AF464764.1 | AAL69962.1 | |||
AK000517.1 | BAA91223.1 | |||
AK000784.1 | BAA91377.1 | |||
AK025952.1 | BAB15293.1 | |||
AK223253.1 | BAD96973.1 | |||
AK223269.1 | BAD96989.1 | |||
AK292681.1 | ||||
AK302989.1 | ||||
BC001039.2 | AAH01039.1 | |||
BC003592.1 | AAH03592.1 | |||
BC039269.1 | AAH39269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3644827 | Chr.19:54976334 - 55017364 on Build GRCh38 | Loss | NLRP2 GP6 |
esv2667609 | Chr.19:54472514 - 55094226 on Build GRCh38 | Deletion | NLRP7 KIR3DX1 LILRP2 NLRP2 LAIR2 FCAR PPP1R12C LOC102724300 NCR1 KIR2DL4 KIR2DS4 EPS8L1 LILRA2 MIR8061 LILRA1 LILRB1 KIR3DL1 GP6 LOC101928804 KIR2DL1 KIR3DL3 KIR3DL2 KIR2DL3 LILRB4 CDC42EP5 RDH13 |
dgv3679n100 | Chr.19:54944244 - 55017462 on Build GRCh38 | Loss | NLRP7 NLRP2 GP6 |
nsv580341 | Chr.19:54973805 - 55048372 on Build GRCh38 | Loss | NLRP2 GP6 RDH13 |
nsv1062047 | Chr.19:54921092 - 55041328 on Build GRCh38 | Gain | NLRP7 NLRP2 NCR1 GP6 RDH13 |
esv3583572 | Chr.19:54937457 - 54998307 on Build GRCh38 | Loss | NLRP7 NLRP2 NCR1 |
esv2659880 | Chr.19:54976502 - 55017273 on Build GRCh38 | Deletion | NLRP2 GP6 |
esv3583573 | Chr.19:54992924 - 55013445 on Build GRCh38 | Loss | NLRP2 |
nsv833880 | Chr.19:54889952 - 55067343 on Build GRCh38 | Loss | NLRP7 NLRP2 NCR1 GP6 FCAR RDH13 |
esv2718872 | Chr.19:54472594 - 55094247 on Build GRCh38 | Deletion | NLRP7 KIR3DX1 LILRP2 NLRP2 LAIR2 FCAR PPP1R12C LOC102724300 NCR1 KIR2DL4 KIR2DS4 EPS8L1 LILRA2 MIR8061 LILRA1 LILRB1 KIR3DL1 GP6 LOC101928804 KIR2DL1 KIR3DL3 KIR3DL2 KIR2DL3 LILRB4 CDC42EP5 RDH13 |
esv3893250 | Chr.19:54906374 - 55000768 on Build GRCh38 | Loss | NLRP7 NLRP2 NCR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |