Assay Details
Target Gene Details
Entrez Gene ID: | 105372441 |
Gene Name: | uncharacterized LOC105372441 |
Gene Aliases: |
KLK3e |
Location: |
Chr.19:50830530-50851143 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105372441 | NR_131203.1 | |||
NR_131205.1 | ||||
AA565890.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3583464 | Chr.19:50818135 - 50842312 on Build GRCh38 | Loss | LOC105372441 KLK15 KLK1 MGC45922 |
nsv2531 | Chr.19:50830372 - 50875341 on Build GRCh38 | Deletion | LOC105372441 KLK3 KLK2 KLK15 |
dgv1058e212 | Chr.19:50828652 - 50842312 on Build GRCh38 | Loss | LOC105372441 KLK15 |