Assay Details
Target Gene Details
Entrez Gene ID: | 4784 |
Gene Name: | nuclear factor I X |
Gene Aliases: |
MRSHSS, NF1A, SOTOS2 |
Location: |
Chr.19:12995512-13098796 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFIX | NM_002501.3 | NP_002492.2 | ||
XM_005259918.4 | XP_005259975.1 | |||
XM_017026837.1 | XP_016882326.1 | |||
BF115883.1 | ||||
BT019732.1 | AAV38537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1055900 | Chr.19:12979147 - 13034197 on Build GRCh38 | Gain | NFIX |
nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion | BEST2 ASNA1 MIR6515 DNASE2 SNORD41 MIR5695 HOOK2 GCDH LOC105372280 RAD23A RTBDN KLF1 DHPS MIR5684 WDR83OS CALR FBXW9 NFIX DAND5 PRDX2 RNASEH2A MIR6794 MAST1 C19orf43 SYCE2 TNPO2 JUNB GADD45GIP1 WDR83 FARSA SNORD135 |
nsv2422 | Chr.19:13000362 - 13032711 on Build GRCh38 | Insertion | NFIX |
nsv833754 | Chr.19:12876208 - 13028186 on Build GRCh38 | Loss | GCDH CALR RAD23A KLF1 MIR6515 SYCE2 DNASE2 GADD45GIP1 NFIX FARSA MIR5695 DAND5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |