Assay Details
Target Gene Details
Entrez Gene ID: | 10055 |
Gene Name: | SUMO1 activating enzyme subunit 1 |
Gene Aliases: |
AOS1, HSPC140, SUA1, UBLE1A |
Location: |
Chr.19:47130782-47210636 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SAE1 | NM_001145713.1 | NP_001139185.1 | ||
NM_001145714.1 | NP_001139186.1 | |||
NM_005500.2 | NP_005491.1 | |||
NR_027280.1 | ||||
XM_006722963.2 | XP_006723026.1 | |||
XM_006722965.3 | XP_006723028.1 | |||
XM_017026135.1 | XP_016881624.1 | |||
XM_017026136.1 | XP_016881625.1 | |||
AF046025.2 | AAD23902.2 | |||
AF090385.2 | AAD12785.2 | |||
AF110956.1 | AAD24433.1 | |||
AF161489.1 | AAF29104.1 | |||
AK021978.1 | ||||
AK027595.1 | ||||
AK055481.1 | ||||
AK090645.1 | ||||
AK302287.1 | ||||
AK315624.1 | ||||
BC000344.2 | AAH00344.1 | |||
BC003611.2 | AAH03611.1 | |||
BC018271.1 | AAH18271.1 | |||
BT007290.1 | AAP35954.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv833852 | Chr.19:47134590 - 47307834 on Build GRCh38 | Loss | MIR3190 MIR3191 INAFM1 SAE1 BBC3 CCDC9 |
nsv833851 | Chr.19:47058544 - 47208779 on Build GRCh38 | Loss | SAE1 ZC3H4 |
nsv1067417 | Chr.19:47103045 - 47309908 on Build GRCh38 | Gain | MIR3190 MIR3191 INAFM1 SAE1 BBC3 ZC3H4 CCDC9 C5AR1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |