Assay Details
Target Gene Details
Entrez Gene ID: | 22900 |
Gene Name: | caspase recruitment domain family member 8 |
Gene Aliases: |
CARDINAL, DACAR, DAKAR, NDPP, NDPP1, TUCAN |
Location: |
Chr.19:48203148-48256269 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv3613n100 | Chr.19:48220413 - 48256972 on Build GRCh38 | Gain | CARD8-AS1 CARD8 |
nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss | CARD8-AS1 NOSIP GFY KDELR1 PRMT1 KCNA7 ZNF473 NUCB1-AS1 SNAR-A12 TEAD2 SPHK2 C19orf68 SNAR-C3 CCDC155 SNAR-A10 SNAR-A13 CGB3 SNAR-A5 SNAR-B1 MIR4749 PLA2G4C-AS1 LHB SNAR-C1 SNAR-A7 RCN3 CD37 TRPM4 AKT1S1 CGB2 GRIN2D TBC1D17 NUCB1 PLEKHA4 IL4I1 TMEM143 RASIP1 KCNJ14 FUT2 LOC105372430 SNAR-G2 LMTK3 CGB1 RUVBL2 PLA2G4C PRRG2 SNORD33 SNORD35B SNAR-A14 PPFIA3 MIR6798 SNORD35A CGB8 FLT3LG DBP SNAR-G1 IRF3 AP2A1 PTOV1-AS2 NTF4 HSD17B14 SNORD32A LIG1 FCGRT PRR12 SNAR-A11 NUP62 SIGLEC16 SNAR-A2 LOC100287477 SNAR-B2 SLC17A7 FUT1 TULP2 FTL MIR6800 ADM5 CGB5 RPS11 MAMSTR CGB7 BCAT2 MIR150 ATF5 PTOV1-AS1 PIH1D1 FGF21 CPT1C CCDC114 SCAF1 HRC PPP1R15A MIR4751 PTOV1 SNAR-C4 GYS1 LIN7B BCL2L12 SYNGR4 EMP3 SNAR-A8 SNAR-A4 PNKP LOC105447645 IZUMO1 ZNF114 MIR4324 SNAR-A9 DHDH PTH2 MIR6799 CABP5 FUZ BSPH1 VRK3 MIR5088 MED25 FLJ26850 SLC6A16 SNRNP70 LOC101928295 LOC101059948 CARD8 TSKS SNAR-C2 FAM83E ALDH16A1 RRAS CYTH2 SIGLEC11 SNAR-D DKKL1 RPL13A MIR4750 SNAR-C5 C19orf73 SPACA4 ELSPBP1 SNAR-A1 SNAR-A3 SEC1P BAX SULT2B1 GRWD1 CA11 SNAR-A6 SNORD34 NTN5 RPL18 |
dgv3614n100 | Chr.19:48224334 - 48283199 on Build GRCh38 | Gain | CARD8-AS1 CARD8 ZNF114 |
nsv469623 | Chr.19:48209728 - 48360236 on Build GRCh38 | Loss | CARD8-AS1 EMP3 TMEM143 CARD8 ZNF114 CCDC114 |
esv34181 | Chr.19:48009377 - 48416682 on Build GRCh38 | Loss | CARD8-AS1 PLA2G4C-AS1 KDELR1 CABP5 ELSPBP1 SYNGR4 PLA2G4C C19orf68 LIG1 GRIN2D EMP3 TMEM143 CARD8 ZNF114 CCDC114 |
nsv579902 | Chr.19:48208219 - 48239130 on Build GRCh38 | Loss | CARD8 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |