Assay Details
Target Gene Details
Entrez Gene ID: | 126328 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit A11 |
Gene Aliases: |
B14.7, CI-B14.7 |
Location: |
Chr.19:5891276-5904013 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NDUFA11 | NM_001193375.1 | NP_001180304.1 | ||
NM_175614.4 | NP_783313.1 | |||
NR_034166.2 | ||||
AJ539081.1 | CAD62165.1 | |||
AK127692.1 | BAC87088.1 | |||
AL546175.3 | ||||
AY726561.1 | ||||
BC069045.1 | AAH69045.1 | |||
BQ054257.1 | ||||
BQ671956.1 | ||||
BQ955016.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)