Assay Details
Target Gene Details
Entrez Gene ID: | 4784 |
Gene Name: | nuclear factor I X |
Gene Aliases: |
MRSHSS, NF1A, SOTOS2 |
Location: |
Chr.19:12995512-13098796 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFIX | NM_002501.3 | NP_002492.2 | ||
XM_005259918.4 | XP_005259975.1 | |||
XM_017026837.1 | XP_016882326.1 | |||
BF115883.1 | ||||
BT019732.1 | AAV38537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1055900 | Chr.19:12979147 - 13034197 on Build GRCh38 | Gain | NFIX |
nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion | RTBDN RNASEH2A TNPO2 BEST2 MAST1 MIR6515 NFIX MIR5684 FBXW9 SYCE2 DHPS MIR6794 WDR83OS RAD23A FARSA JUNB SNORD135 LOC105372280 PRDX2 MIR5695 DAND5 HOOK2 DNASE2 C19orf43 SNORD41 KLF1 ASNA1 CALR GCDH WDR83 GADD45GIP1 |
nsv2422 | Chr.19:13000362 - 13032711 on Build GRCh38 | Insertion | NFIX |
nsv833754 | Chr.19:12876208 - 13028186 on Build GRCh38 | Loss | KLF1 NFIX SYCE2 MIR5695 CALR RAD23A FARSA DAND5 GCDH MIR6515 GADD45GIP1 DNASE2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |