Assay Details
Target Gene Details
Entrez Gene ID: | 83639 |
Gene Name: | testis expressed 101 |
Gene Aliases: |
CT131, GTPR867, NYD-SP8, PRO1884, SGRG, SPATA44, TES101RP |
Location: |
Chr.19:43388611-43418615 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TEX101 | NM_031451.4 | NP_113639.4 | ||
AF241268.1 | AAK28327.1 | |||
BC001861.2 | AAH01861.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv469666 | Chr.19:43272995 - 43430182 on Build GRCh38 | Loss | CD177 PRG1 TEX101 |
nsv1057949 | Chr.19:43380906 - 43431018 on Build GRCh38 | Loss | TEX101 |
esv3644466 | Chr.19:43383591 - 43418114 on Build GRCh38 | Gain | TEX101 |
dgv3603n100 | Chr.19:43304322 - 43590925 on Build GRCh38 | Gain | ETHE1 CD177 LYPD3 PINLYP PRG1 PHLDB3 ZNF575 TEX101 IRGQ XRCC1 |
nsv1060280 | Chr.19:43362671 - 43455533 on Build GRCh38 | Gain | CD177 TEX101 |
nsv1057657 | Chr.19:42798345 - 43572412 on Build GRCh38 | Gain | PSG11 LOC284344 PSG2 PSG7 PSG1 PSG10P ZNF575 TEX101 XRCC1 ETHE1 CD177 LYPD3 PSG6 PRG1 PHLDB3 PSG5 PSG4 LOC100289650 PSG9 |
nsv520852 | Chr.19:43380933 - 43455962 on Build GRCh38 | Gain+Loss | TEX101 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |