Assay Details
Target Gene Details
Entrez Gene ID: | 23640 |
Gene Name: | HSPA (Hsp70) binding protein 1 |
Gene Aliases: |
FES1 |
Location: |
Chr.19:55262222-55280383 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSPBP1 | NM_001130106.1 | NP_001123578.1 | ||
NM_001297600.1 | NP_001284529.1 | |||
NM_012267.4 | NP_036399.3 | |||
XM_005258700.2 | XP_005258757.1 | |||
XM_005258701.2 | XP_005258758.1 | |||
XM_011526691.1 | XP_011524993.1 | |||
XM_017026543.1 | XP_016882032.1 | |||
XM_017026544.1 | XP_016882033.1 | |||
XM_017026545.1 | XP_016882034.1 | |||
AB020592.1 | BAB18742.1 | |||
AF093420.1 | AAC79703.1 | |||
AF187859.1 | AAF35833.1 | |||
AF217996.1 | AAG17238.1 | |||
AK075293.1 | ||||
AK129755.1 | ||||
AK130781.1 | ||||
AK294358.1 | ||||
BC001236.1 | AAH01236.1 | |||
BC002373.2 | AAH02373.1 | |||
CR457118.1 | CAG33399.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |