Assay Details
Target Gene Details
Entrez Gene ID: | 4784 |
Gene Name: | nuclear factor I X |
Gene Aliases: |
MRSHSS, NF1A, SOTOS2 |
Location: |
Chr.19:12995512-13098796 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFIX | NM_002501.3 | NP_002492.2 | ||
XM_005259918.4 | XP_005259975.1 | |||
XM_017026837.1 | XP_016882326.1 | |||
BF115883.1 | ||||
BT019732.1 | AAV38537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1055900 | Chr.19:12979147 - 13034197 on Build GRCh38 | Gain | NFIX |
nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion | MIR6794 WDR83 WDR83OS CALR SYCE2 MAST1 ASNA1 LOC105372280 KLF1 RTBDN MIR6515 PRDX2 MIR5684 TNPO2 MIR5695 SNORD41 RNASEH2A C19orf43 DHPS FARSA SNORD135 JUNB GCDH GADD45GIP1 HOOK2 DNASE2 FBXW9 RAD23A NFIX DAND5 BEST2 |
nsv2422 | Chr.19:13000362 - 13032711 on Build GRCh38 | Insertion | NFIX |
nsv833754 | Chr.19:12876208 - 13028186 on Build GRCh38 | Loss | DNASE2 CALR SYCE2 RAD23A FARSA NFIX DAND5 KLF1 GCDH MIR6515 MIR5695 GADD45GIP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |