Assay Details
Target Gene Details
Entrez Gene ID: | 9524 |
Gene Name: | trans-2,3-enoyl-CoA reductase |
Gene Aliases: |
GPSN2, MRT14, SC2, TER |
Location: |
Chr.19:14527760-14565980 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TECR | NM_001321170.1 | NP_001308099.1 | ||
NM_138501.5 | NP_612510.1 | |||
NR_038103.1 | ||||
NR_038104.1 | ||||
XM_006722945.1 | XP_006723008.1 | |||
XM_011528442.2 | XP_011526744.1 | |||
AF038958.1 | AAC39872.1 | |||
AF038959.1 | AAC39873.1 | |||
AF222742.1 | AAF32373.1 | |||
AK001416.1 | ||||
AK094087.1 | ||||
AK299134.1 | ||||
AK315412.1 | ||||
BC002425.1 | AAH02425.1 | |||
BC005952.1 | AAH05952.1 | |||
BC007801.2 | AAH07801.1 | |||
BC013881.2 | AAH13881.1 | |||
BG705196.1 | ||||
DB491783.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs142364787] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |