Assay Details
Target Gene Details
Entrez Gene ID: | 4784 |
Gene Name: | nuclear factor I X |
Gene Aliases: |
MRSHSS, NF1A, SOTOS2 |
Location: |
Chr.19:12995512-13098796 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NFIX | NM_002501.3 | NP_002492.2 | ||
XM_005259918.4 | XP_005259975.1 | |||
XM_017026837.1 | XP_016882326.1 | |||
BF115883.1 | ||||
BT019732.1 | AAV38537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv828444 | Chr.19:13012681 - 13027353 on Build GRCh38 | Loss | NFIX |
nsv1055900 | Chr.19:12979147 - 13034197 on Build GRCh38 | Gain | NFIX |
nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion | MIR6515 WDR83OS DHPS MIR5695 PRDX2 SNORD41 NFIX JUNB RTBDN LOC105372280 MIR5684 GADD45GIP1 SNORD135 SYCE2 KLF1 HOOK2 GCDH MIR6794 WDR83 BEST2 FARSA DAND5 FBXW9 RAD23A RNASEH2A DNASE2 CALR MAST1 ASNA1 C19orf43 TNPO2 |
nsv2422 | Chr.19:13000362 - 13032711 on Build GRCh38 | Insertion | NFIX |
nsv833754 | Chr.19:12876208 - 13028186 on Build GRCh38 | Loss | MIR6515 DNASE2 MIR5695 FARSA NFIX CALR SYCE2 DAND5 KLF1 RAD23A GADD45GIP1 GCDH |
esv275430 | Chr.19:13015145 - 13020565 on Build GRCh38 | Gain+Loss | NFIX |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |