Assay Details
Target Gene Details
Entrez Gene ID: | 55640 |
Gene Name: | feline leukemia virus subgroup C cellular receptor family member 2 |
Gene Aliases: |
C14orf58, CCT, EPV, FLVCRL14q, MFSD7C, PVHH |
Location: |
Chr.14:75578597-75648169 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FLVCR2 | NM_001195283.1 | NP_001182212.1 | ||
NM_017791.2 | NP_060261.2 | |||
AF456126.1 | AAO15528.1 | |||
AK000378.1 | BAA91126.1 | |||
AK297002.1 | ||||
AY260572.1 | AAP86633.1 | |||
AY260577.1 | AAP86638.1 | |||
BC019087.2 | AAH19087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1037822 | Chr.14:75468606 - 75633843 on Build GRCh38 | Gain | MIR7641-2 LOC102724153 FLVCR2 JDP2 BATF |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |