Assay Details
Target Gene Details
Entrez Gene ID: | 7051 |
Gene Name: | transglutaminase 1 |
Gene Aliases: |
ARCI1, ICR2, KTG, LI, LI1, TGASE, TGK |
Location: |
Chr.14:24249114-24263210 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TGM1 | NM_000359.2 | NP_000350.1 | ||
AK291350.1 | ||||
AK299436.1 | ||||
AK301652.1 | ||||
AK315819.1 | ||||
AK315828.1 | ||||
AK315843.1 | ||||
BC034699.1 | AAH34699.1 | |||
D90287.1 | BAA14329.1 | |||
M55183.1 | AAA59474.1 | |||
M62925.1 | AAA61166.1 | |||
X57974.1 | CAA41040.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv952818 | Chr.14:24249495 - 24270894 on Build GRCh38 | Deletion | RABGGTA TGM1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |