Assay Details
Target Gene Details
Entrez Gene ID: | 145264 |
Gene Name: | serpin family A member 12 |
Gene Aliases: |
OL-64 |
Location: |
Chr.14:94481651-94517844 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERPINA12 | XM_011536455.1 | XP_011534757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832863 | Chr.14:94348482 - 94501487 on Build GRCh38 | Gain | SERPINA1 SERPINA12 SERPINA2 SERPINA11 SERPINA9 |
nsv510649 | Chr.14:94421103 - 94544514 on Build GRCh38 | Deletion | SERPINA12 SERPINA11 SERPINA9 |
nsv470662 | Chr.14:94296542 - 94798793 on Build GRCh38 | Gain | SERPINA5 GSC SERPINA1 SERPINA6 SERPINA13P SERPINA3 SERPINA4 SERPINA12 SERPINA2 SERPINA11 SERPINA9 |
nsv565623 | Chr.14:94393116 - 94635006 on Build GRCh38 | Gain | SERPINA5 SERPINA3 SERPINA4 SERPINA12 SERPINA11 SERPINA9 |
nsv832864 | Chr.14:94362101 - 94538150 on Build GRCh38 | Loss | SERPINA1 SERPINA12 SERPINA2 SERPINA11 SERPINA9 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |