Assay Details
Target Gene Details
Entrez Gene ID: | 3831 |
Gene Name: | kinesin light chain 1 |
Gene Aliases: |
KLC, KNS2, KNS2A |
Location: |
Chr.14:103629188-103701551 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KLC1 | NM_001130107.1 | NP_001123579.1 | ||
NM_005552.4 | NP_005543.2 | |||
NM_182923.3 | NP_891553.2 | |||
AF527736.1 | AAP85635.1 | |||
AK294508.1 | ||||
AY180164.2 | AAO62549.1 | |||
BC008881.2 | AAH08881.1 | |||
BP274336.1 | ||||
DA501532.1 | ||||
L04733.1 | AAA16576.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635603 | Chr.14:103624778 - 103714856 on Build GRCh38 | Gain | XRCC3 KLC1 LOC107984648 |
nsv832883 | Chr.14:103461350 - 103654616 on Build GRCh38 | Loss | CKB MARK3 TRMT61A BAG5 KLC1 APOPT1 |
nsv566023 | Chr.14:103595309 - 103731914 on Build GRCh38 | Loss | ZFYVE21 XRCC3 KLC1 LOC107984648 |
nsv1054574 | Chr.14:103210111 - 105365114 on Build GRCh38 | Loss | C14orf2 MIR203B ZBTB42 PACS2 AKT1 EIF5 LOC101929634 LINC00638 NUDT14 RD3L KIF26A AHNAK2 PLD4 TMEM179 MIR203A LOC105378183 MIR6765 C14orf180 PPP1R13B LOC105370708 LOC102723354 LINC00637 BTBD6 ZFYVE21 INF2 CDCA4 MIR4710 SIVA1 MARK3 BRF1 TRMT61A ADSSL1 LOC388022 XRCC3 KLC1 LOC107984648 C14orf79 APOPT1 CEP170B JAG2 TDRD9 ASPG SNORA28 CKB LOC105370687 LOC105370691 GPR132 BAG5 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |