Assay Details
Target Gene Details
Entrez Gene ID: | 1033 |
Gene Name: | cyclin dependent kinase inhibitor 3 |
Gene Aliases: |
CDI1, CIP2, KAP, KAP1 |
Location: |
Chr.14:54396939-54420218 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDKN3 | NM_001130851.1 | NP_001124323.1 | ||
NM_005192.3 | NP_005183.2 | |||
XM_011536353.2 | XP_011534655.1 | |||
AK098653.1 | ||||
AK293265.1 | ||||
BC064965.1 | AAH64965.1 | |||
BC103633.1 | AAI03634.1 | |||
EF560750.1 | ||||
KP966095.1 | ||||
L25876.1 | AAA60222.1 | |||
L27711.1 | AAA66496.1 | |||
U02681.1 | AAC04932.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv832801 | Chr.14:54255297 - 54420055 on Build GRCh38 | Gain | CDKN3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |