Assay Details
Target Gene Details
Entrez Gene ID: | 5104 |
Gene Name: | serpin family A member 5 |
Gene Aliases: |
PAI-3, PAI3, PCI, PCI-B, PLANH3, PROCI |
Location: |
Chr.14:94581369-94593120 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SERPINA5 | NM_000624.5 | NP_000615.3 | ||
AK096131.1 | ||||
AK293192.1 | ||||
AK298279.1 | ||||
AK310869.1 | ||||
BC008915.2 | AAH08915.1 | |||
DC394575.1 | ||||
HY003178.1 | ||||
J02639.1 | AAA35688.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv565626 | Chr.14:94583971 - 94592873 on Build GRCh38 | Loss |
![]() |
nsv565625 | Chr.14:94567019 - 94603850 on Build GRCh38 | Loss |
![]() ![]() |
nsv470662 | Chr.14:94296542 - 94798793 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv565623 | Chr.14:94393116 - 94635006 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv832865 | Chr.14:94499457 - 94667894 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv3635384 | Chr.14:94569665 - 94707857 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)