Assay Details
Target Gene Details
Entrez Gene ID: | 8814 |
Gene Name: | cyclin dependent kinase like 1 |
Gene Aliases: |
KKIALRE, P42 |
Location: |
Chr.14:50329404-50397404 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDKL1 | NM_001282236.1 | NP_001269165.1 | ||
NM_004196.4 | NP_004187.2 | |||
XM_005268157.2 | XP_005268214.1 | |||
XM_005268159.2 | XP_005268216.1 | |||
XM_005268160.4 | XP_005268217.1 | |||
XM_011537276.1 | XP_011535578.1 | |||
XM_017021729.1 | XP_016877218.1 | |||
XM_017021730.1 | XP_016877219.1 | |||
XM_017021731.1 | XP_016877220.1 | |||
XM_017021732.1 | XP_016877221.1 | |||
AA780354.1 | ||||
AK308732.1 | ||||
BC104977.1 | AAI04978.1 | |||
X66358.1 | CAA47002.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv564835 | Chr.14:50024482 - 50439753 on Build GRCh38 | Gain | MAP4K5 MIR4504 VCPKMT LINC01599 CDKL1 ATP5S SOS2 L2HGDH |
nsv832793 | Chr.14:50294245 - 50470857 on Build GRCh38 | Loss | MAP4K5 MIR4504 CDKL1 ATP5S L2HGDH |
nsv1052059 | Chr.14:50291535 - 50364621 on Build GRCh38 | Loss | MIR4504 CDKL1 ATP5S L2HGDH |
esv28926 | Chr.14:49426970 - 50475809 on Build GRCh38 | Gain+Loss | KLHDC2 POLE2 RN7SL2 LRR1 VCPKMT RPS29 ARF6 MGAT2 LOC100288910 DNAAF2 SOS2 LOC100506446 RPL36AL MAP4K5 MIR4504 LINC01588 KLHDC1 LINC01599 CDKL1 NEMF ATP5S RN7SL1 MIR6076 L2HGDH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |