Assay Details
Target Gene Details
Entrez Gene ID: | 64067 |
Gene Name: | neuronal PAS domain protein 3 |
Gene Aliases: |
MOP6, PASD6, bHLHe12 |
Location: |
Chr.14:32934785-33804176 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |