Assay Details
Target Gene Details
Entrez Gene ID: | 10538 |
Gene Name: | basic leucine zipper ATF-like transcription factor |
Gene Aliases: |
B-ATF, BATF1, SFA-2, SFA2 |
Location: |
Chr.14:75522441-75546992 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BATF | NM_006399.3 | NP_006390.1 | ||
BC032294.1 | AAH32294.1 | |||
D42106.1 | BAA07686.1 | |||
U15460.1 | AAC50314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1037822 | Chr.14:75468606 - 75633843 on Build GRCh38 | Gain | JDP2 FLVCR2 BATF MIR7641-2 LOC102724153 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |