Assay Details
Target Gene Details
Entrez Gene ID: | 105370705 |
Gene Name: | collagen alpha-2(I) chain-like |
Gene Aliases: |
- |
Location: |
Chr.14:23056023-23058476 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC105370705 | XM_011537450.1 | XP_011535752.1 |