Assay Details
Target Gene Details
Entrez Gene ID: | 5587 |
Gene Name: | protein kinase D1 |
Gene Aliases: |
PKC-MU, PKCM, PKD, PRKCM |
Location: |
Chr.14:29576479-29927693 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PRKD1 | NM_002742.2 | NP_002733.2 | ||
XM_005267859.1 | XP_005267916.1 | |||
XM_011536964.2 | XP_011535266.1 | |||
XM_011536965.2 | XP_011535267.1 | |||
XM_017021461.1 | XP_016876950.1 | |||
XM_017021462.1 | XP_016876951.1 | |||
AK314170.1 | ||||
X75756.1 | CAA53384.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv526536 | Chr.14:28322483 - 30669172 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv564160 | Chr.14:29030900 - 29631166 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)