Assay Details
Target Gene Details
Entrez Gene ID: | 122945 |
Gene Name: | NADP dependent oxidoreductase domain containing 1 |
Gene Aliases: |
C14orf148 |
Location: |
Chr.14:77394021-77426013 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NOXRED1 | NM_001113475.2 | NP_001106946.1 | ||
XM_005267330.3 | XP_005267387.1 | |||
XM_011536427.2 | XP_011534729.1 | |||
XM_011536428.2 | XP_011534730.1 | |||
XM_011536429.2 | XP_011534731.1 | |||
XM_017020969.1 | XP_016876458.1 | |||
XM_017020970.1 | XP_016876459.1 | |||
AK057371.1 | ||||
BC066965.1 | AAH66965.1 | |||
BI829015.1 | ||||
DB021944.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1362 | Chr.14:77386859 - 77432262 on Build GRCh38 | Deletion | SAMD15 VIPAS39 NOXRED1 |
nsv85801 | Chr.14:77408913 - 77409813 on Build GRCh38 | Deletion | NOXRED1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |